Learn More
Wesbart™ Side Access Freezer Rack
Click to view available options
Holds:
Boxes (16)
Boxes (20)
Boxes (24)
Dimensions (L x W x H):
560 x 140 x 235 mm
560 x 140 x 339 mm
565 x 140 x 280 mm
Unit Size:
Each
Description
ULT freezer racks for upright freezers to store boxes
Specifications
Specifications
| Product Type | Freezer Rack |
| Description | Aluminum Side Access Rack |
| Autoclavable | Autoclavable |
| Color | Silver |
| Holds | Boxes (16) |
| Shape | Rectangular |
| Stackable | Yes |
| Sterility | Non-sterile |
| Dimensions (L x W x H) | 560 x 140 x 235 mm |
| Height (Metric) | 235 mm |