Learn More
Wesbart™ Side Access Freezer Rack
Product Code. 16601246
Click to view available options
Holds:
Boxes (16)
Boxes (20)
Boxes (24)
Dimensions (L x W x H):
560 x 140 x 235 mm
560 x 140 x 339 mm
565 x 140 x 280 mm
Unit Size:
Each
Product Code. 16601246
Brand: Wesbart™ SOR196
Only null left
Add to basket
Add to basket
Wesbart side loading racks to store cryoboxes.
ULT freezer racks for upright freezers to store boxes
Specifications
Product Type Freezer Rack
Description Aluminum Side Access Rack
Autoclavable Autoclavable
Color Silver
Holds Boxes (16)
Shape Rectangular
Stackable Yes
Sterility Non-sterile
Dimensions (L x W x H) 560 x 140 x 235 mm
Height (Metric) 235 mm