Learn More
Sibutramine hydrochloride monohydrate
Product Code. 15781799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15781799 5 g 5g
15771799 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15781799 Supplier Thermo Scientific Acros Supplier No. 462200050
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 125494-59-9 | C17H29Cl2NO | 334.33 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 125494-59-9
Molecular Formula C17H29Cl2NO
Molecular Weight (g/mol) 334.33
InChI Key KFNNPQDSPLWLCX-UHFFFAOYNA-N
PubChem CID 64765
Specifications
Quantity 5 g
Chemical Name or Material Sibutramine hydrochloride monohydrate