missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sibofimloc, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
Sibofimloc (Antibiotic-202) is a first-in-class, gut-restricted, orally active FimH adhesion inhibitor extracted from patent WO2014100158A1, Compound Example 202. Sibofimloc has anti-bacterial infective activity. Sibofimloc is developed for inflammatory bowel disease (IBD).
Specifications
Specifications
| Chemical Name or Material | Sibofimloc |
| Physical Form | Powder |
| CAS | 1616113-45-1 |
| Color | Off-White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C35H39NO11 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Antibiotic-202 |
| Solubility Information | DMSO : ≥ 31 mg/mL (47.72 mM) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction