Learn More
SHOWA S-TEX KV3 Cut-Resistant Knit Gloves
Beschreibung
- The glove has a natural rubber palm dip that ensures a superior grip in both dry and wet conditions.
- For maximum cut resistance, the KV3 glove is made with a combination of stainless steel liner, Kevlar fibers, and patented Hagane Coil technology.
- The KV3 glove is designed to be form-fitting and breathable, ensuring maximum comfort and performance during use.
Spezifikation
Spezifikation
| Construction | Seamless Knit |
| Coating Material | Latex |
| Color | Black, Yellow |
| Hand | Specific |
| Latex Free | No |
| Material | Aramid Knit, Polyester, Stainless Steel Fiber |
| Product Type | Cut-resistant Gloves |
| Size | Medium |
| Gender | Unisex |
| Glove Cuff Style | Knit |
| Mehr anzeigen |