Learn More
SHOWA™ Blue Nitrile N-Dex Medical Exam Gloves
Product Code. 10014331
Click to view available options
Size:
Large+
Small
X-Large
Unit Size:
Pack of 100
Pack of 50
This item is not returnable. View return policy
Product Code. 10014331
Brand: SHOWA™ G9905PFS
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This item is not returnable. View return policy
Protect hands with SHOWA™ Blue Nitrile N-Dex Medical Exam Gloves since they provide three times the puncture resistance of natural rubber or vinyl gloves of comparable thickness. .
  • Powder-free
  • Rolled cuff
  • Blue
  • Disposable
Specifications
Color Blue
Disposable Single-use
Material Nitrile
Size Small