missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ 6797R Chemical Resistant Neoprene Gloves, Extended Sleeve
Description
- Extended-arm protection
- Protection against acids, caustics, oils, greases and many solvents
- Protection against physical hazards such as abrasion and cut
- Cotton liner is cool and comfortable in hot conditions and keeps wearer warm in cold conditions
- Ideal applications include:
- Chemical spray and treatment
- Coating preparation
- Laboratory, pharma and analysis
- Painting and spray workshops
Specifications
Specifications
| Certifications/Compliance | 21 CFR 170-199 |
| Certifications/Compliance (Europe) | Cat III CE 0321, EN 407:3242XX |
| Applications | Laboratory |
| Color | Black |
| Latex Free | Yes |
| Material | Neoprene |
| Product Type | Neoprene Gloves |
| Size | 10 |
| EN388 Standard | EN 388:3111X |
| EN 421 | No |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?