Learn More
SHIELD Scientific SHIELDskin CHEM™ NEO NITRILE™ 300
Product Code. 17862244
Encompass_Preferred
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 400
Product Code. 17862244
Brand: SHIELD Scientific 66 9251_10
Only null left
Add to basket
Add to basket
SHIELDskin CHEM™ gloves mark a new level of comfort and protection. SHIELDskin CHEM™ gloves have been designed for laboratory use to protect you from chemical hazards. Using a new CR/NBR blend and the double-dipping process, they protect you from hazards, without compromising comfort.
  • Brand signature: Red for chemical risk
  • Laboratory gloves for chemical protection
  • Neoprene/nitrile gloves
  • Non-sterile gloves
  • Maximum comfort
  • Compliance with the latest European standards
  • Category III PPE gloves (Regulation (EU) 2016/425)
  • Class 1 MDR (Regulation (EU) 2017/745)
  • Extra length from 300 mm (exceeds ISO 21420:2020+A1:2022)
  • Biohazard: ISO 374-5:2016
  • Biosafety: AQL 0.25 (ISO 374-2:2019 Level  3)
  • Virus resistant (ISO 16604:2004 procedure B)
  • Extensively tested for chemical permeation according to ISO 374-1:2016+A1:2018 (Test method EN 16523-1:2015+A1:2018)
  • Reduced risk of allergies
  • Double-walled protection afforded by the twinSHIELD™ technology
  • Packaging designed to comply with laboratory environments processes
Specifications
Certifications/Compliance AQL 0.25
Applications Laboratory, Industrial
Color Red
Finger Thickness (English) 15.7 mil
Finger Thickness (Metric) 0.40 mm
Latex Free Yes
Material Nitrile, Neoprene
Palm Thickness (English) 12.2 mil
Palm Thickness (Metric) 0.31 mm
Size X-Small
Thickness (English) 7.9 mil
Thickness (Metric) 0.20 mm
Glove Cuff Style Beaded