missing translation for 'onlineSavingsMsg'
Learn More

Sheep Anti-Mouse IgG H&L (TRITC), Abcam

Product Code. 32116331
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32116331 1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32116331 Supplier Abcam Supplier No. ab68051mg
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Sheep Anti-Mouse IgG H&L (TRITC)

The product is subject to the following: Abcam Restricted Use Statement

.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Antigen Ighg1
Applications ELISA, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy, Western Blot
Cellular Localization Isoform Secreted: Secreted
Classification Polyclonal
Conjugate TRITC
Formulation pH: 7.2, Preservative: 0.01% Sodium azide, Constituents: 1% BSA, 0.88% Sodium chloride, 0.424% Potassium phosphate solution
Gene Ighg1
Gene Accession No. P01868
Host Species Sheep
Immunogen Ighg1
Quantity 1 mg
Regulatory Status RUO
Research Discipline Immunology & Infectious Disease
Primary or Secondary Secondary
Target Species Mouse
Content And Storage -20°C
Product Type Primary Antibody
Form Lyophilized
Isotype Not Available
Shipping Condition Blue Ice
Target Gene Ighg1
Target Molecule Ighg1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.