missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fructosyl-Amino Acid Oxidase, Sheep, Polyclonal Antibody, Abnova™
Sheep polyclonal antibody raised against native Fructosyl-Amino Acid Oxidase (E. coli).
Brand: Abnova PAB28098.100ug
Additional Details : Weight : 0.00010kg
Specifications
Fructosyl-Amino Acid Oxidase | |
Polyclonal | |
Sheep polyclonal antibody raised against native Fructosyl-Amino Acid Oxidase (E. coli). | |
Lyophilized from 20mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
Native purified Escherichia coli Fructosyl-Amino Acid Oxidase. | |
RUO | |
Bacteria | |
Lyophilized |
ELISA, Immunoprecipitation, Western Blot | |
Unconjugated | |
ELISA (1:5000 - 1:20000) Immunoprecipitation (1:100) Western Blot (1:500-1:5000) The optimal working dilution should be determined by the end user. | |
Sheep | |
100 μg | |
Primary | |
Store at 4°C on dry atmosphere. After reconstitution with 0.1mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
For Research Use Only