missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC4 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | SHC4 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16870464
|
Proteintech
12641-1-AP-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16860464
|
Proteintech
12641-1-AP-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Activates both Ras-dependent and Ras-independent migratory pathways in melanomas. Contributes to the early phases of agrin-induced tyrosine phosphorylation of CHRNB1.Specifications
| SHC4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q6S5L8, Q6S5L9 | |
| 271849, 399694 | |
| SHC4 Fusion Protein Ag3336 | |
| Primary | |
| -20°C | |
| SHC4 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| hShcD, Rai like protein, RaLP, SH2 domain protein C4, SHC transforming protein 4, SHC transforming protein D, SHC4, SHCD | |
| SHC4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title