missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SHB Recombinant Rabbit Monoclonal Antibody (HL1455)
GREENER_CHOICE

Product Code. 17899505
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17899505 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17899505 Supplier Invitrogen™ Supplier No. MA547053

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The SH2 (Src Homology 2) domain is a structurally conserved motif that contains two alpha helices and seven beta strands and is found in a variety of proteins that are involved in signal transduction throughout the cell. Specifically, the SH2 domain targets SH2 domain-containing proteins to tyrosinephosphorylated sites, an event that can trigger a protein-protein interaction cascade which may ultimately effect gene expression and cellular function. Shb (SH2 domain-containing adapter protein b), Shd (SH2 domain-containing adapter protein d), She (SH2 domain-containing adapter protein e) and Shf(SH2 domain-containing adapter protein f) are SH2 domain-containing proteins that play various roles throughout the cell.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SHB
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL1455
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene SHB
Gene Accession No. Q15464, Q6PD21
Gene Alias bA3J10.2; BC028832; SH2 domain containing adaptor protein B; SH2 domain-containing adapter protein B; Shb; SHB (Src homology 2 domain containing) adaptor protein B; SHB adaptor protein (a Src homology 2 protein); Src homology 2 domain containing adaptor protein B; src homology 2 domain-containing transforming protein B
Gene Symbols SHB
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human SHB. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 230126, 6461
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.