Learn More
Honeywell™ SHARPFLEX PU Gloves
Product Code. 15866735
Change view
Click to view available options
Size:
7
8
9
10
11
Unit Size:
1 pair
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
15866735 11 1 pair
15826735 7 1 pair
15836735 8 1 pair
15846735 9 1 pair
15856735 10 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 15866735 Supplier Honeywell™ Supplier No. 223252311
  • ONLINE EXCLUSIVE PRICE
    11.55 €
    8.85 € / 1 pairs
    Save 2.70 €
    23% Off
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Honeywell™ SHARPFLEX PU Gloves for handling heavy parts with high cut risks in a dry or slightly greasy environment.
  • Knitted glove in composite and para-aramid yarn
  • The knitting technique provides the glove with excellent fit and high dexterity
  • The composite yarn is reliable and safe protection
  • The para-aramid yarn assure a resistance to cut risks
  • Plated polyamide inside
  • Offer a greater comfort and limit the contact of the skin with composite fiber
  • Black PU coating on palm
  • Increase the abrasion resistance and allows to use in slighy greasy environements
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards
Description Para-aramid and composite fiber, 13 gauge
Coating Material Polyurethane
Color Green, Yellow, Black
Size 11
Length (Metric) 25 cm