Learn More
Epredia™ Shandon™ Octospot™ Accessories
Product Code. 12687886 Shop All Epredia Products
Click to view available options
Type:
8-well Strips
8-well Strips, Loose
EZ-Write Slides, 8 spots
Holding Block
Solvent-Resistant Slides, 8 spots
Thick Filter Cards
Unit Size:
Each
Pack of 100
Pack of 5
Pack of 60
Product Code. 12687886
Brand: Epredia™ 1001642
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
These Epredia™ accessories are for use with Shandon Octospot Starter Packs.
TRUSTED_SUSTAINABILITY
Specifications
Type 8-well Strips, Loose
For Use With (Equipment) Shandon™ Octospot™ Starter Packs