Learn More
Epredia™ Small Charcoal Filter
Product Code. 12623106 Shop All Epredia Products
Change view
Click to view available options
Size: 11.25 x 5.75 x 1.31 in. (29 x 15 x 3.3 cm); Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
12623106 Size: 11.25 x 5.75 x 1.31 in. (29 x 15 x 3.3 cm); Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12623106 Supplier Epredia™ Supplier No. 9990610
  • ONLINE EXCLUSIVE PRICE
    186.39 €
    178.29 € / Pack of 1
    Save 8.10 €
    4% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
For use with Epredia™ Hyperclean Pathology Workstations.
Accessories:
  • Charcoal Filter
  • Fine particle Filters
  • Formaldehyde Neutralizing (Potassium Permanganate) Filter
TRUSTED_SUSTAINABILITY
Specifications
Product Type Charcoal Filter
Description Size: 29 cm x 15 cm x 3.3 cm
For Use With (Equipment) Epredia Hyperclean Pathology Workstations, Models 1, 20, 22, 36, 60
Product Line Hyperclean
Quantity 6 Pk.