missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Gram Stain

Product Code. p-9878008 Shop All Epredia Products
Click to view available options
Type:
Gram Crystal Violet stain
Gram Decolorizing Solution
Unit Size:
4 L
This item is not returnable. View return policy

Product Code. 12589956

Brand: Epredia™ 9990725

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Gram crystal violet stain, 4L

Differentiate between Gram-positive and Gram-negative bacteria with Epredia™ Shandon™ Gram Stain.

  • Based upon the popular Hucker modification
  • Demonstrates the presence of microorganisms such as bacteria
TRUSTED_SUSTAINABILITY

Specifications

Type Gram Crystal Violet stain
Item Description Crystal violet gram stain
Quantity 4L
compliance-icons
Product Identifier
  • Shandon Crystal Violet Stain
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, C.I. basicviolet 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.