Learn More
Epredia™ Shandon™ Gram Stain
Shop All Epredia Products
Click to view available options
Type:
Gram Crystal Violet stain
Gram Decolorizing Solution
Unit Size:
4 L
Description
Includes
Gram crystal violet stain, 4L
- Based upon the popular Hucker modification
- Demonstrates the presence of microorganisms such as bacteria
Specifications
Specifications
| Type | Gram Crystal Violet stain |
| Item Description | Crystal violet gram stain |
| Quantity | 4L |
Safety and Handling
Product Identifier
- Shandon Crystal Violet Stain
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- H412-Harmful to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P391-Collect spillage.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : ethanol, C.I. basicviolet 1