Learn More
Epredia™ Shandon™ Gram Stain
Shop All Epredia Products
Click to view available options
Type:
Gram Crystal Violet stain
Gram Decolorizing Solution
Unit Size:
4 L
Description
Includes
Gram decolorizing solution, 4L
- Based upon the popular Hucker modification
- Demonstrates the presence of microorganisms such as bacteria
Specifications
Specifications
| Type | Gram Decolorizing Solution |
| Item Description | Decolorizing Solution |
| Quantity | 4L |