Learn More
Novus Biologicals™ SH3BGRL Overexpression Lysate
Product Code. 18211790 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18211790 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18211790 Supplier Novus Biologicals™ Supplier No. NBL115924
Please to purchase this item. Need a web account? Register with us today!
The SH3BGRL Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Cell Line The lysate was created in HEK293T cells, using plasmid ID RC24993 and based on accession number NM_322. The protein contains a C-terminal DDK tag.
For Use With (Application) Western Blot
Quantity 0.1 mg
Storage Buffer RIPA buffer
Purity Protein
Test Specificity Homo sapiens SH3 domain binding glutamic acid-rich protein like (SH3BGRL), mRNA.
Content And Storage -80°C. Avoid freeze-thaw cycles.
Protein State Native