missing translation for 'onlineSavingsMsg'
Learn More

SGSM1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18494998 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18494998 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18494998 Supplier Novus Biologicals Supplier No. NBP281792AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SGSM1 Polyclonal antibody specifically detects SGSM1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGSM1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias RUN and TBC1 domain-containing protein 2, RUTBC2, small G protein signaling modulator 1, small G protein signaling modulator 1 protein
Host Species Rabbit
Immunogen SGSM1 antibody was raised against a 17 amino acid synthetic peptide near the center of human SGSM1. The immunogen is located within amino acids 480 - 530 of SGSM1. Amino Acid Squence: SVSNLPSLWQPSPRKSS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 129049
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.