missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SGLT2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16678364
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16678364 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16678364 Supplier Invitrogen™ Supplier No. PA5101893

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC5A2.

Sodium-dependent glucose transporter. Has a Na+ to glucose coupling ratio of 1:1. Efficient substrate transport in mammalian kidney is provided by the concerted action of a low affinity high capacity and a high affinity low capacity Na+/glucose cotransporter arranged in series along kidney proximal tubules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SGLT2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC5A2
Gene Accession No. P31639, P53792, Q923I7
Gene Alias glucose transporter; bSGLT2; low affinity Na-dependent glucose transporter (SGLT2); low affinity sodium-dependent glucose cotransporter; low affinity sodium-glucose cotransporter; Na(+)/glucose cotransporter 2; Na(+)/nucleoside cotransporter; Na+/glucose cotransporter 2; Na+/nucleoside cotransporter; Sglt2; SGLT-2; SLC5A2; SNST1; sodium/glucose cotransporter 2; Sodium/nucleoside cotransporter; sodium-glucose cotransporter-like 2; solute carrier family 5 (sodium/glucose cotransporter), member 2; solute carrier family 5 (sodium/glucose transporter), member 2; solute carrier family 5 member 2
Gene Symbols SLC5A2
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC5A2(Accession P31639), corresponding to amino acid residues G110-F160.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246787, 64522, 6524
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.