missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SGK (SGK1), GST Tag Recombinant Protein
Human SGK (SGK1), GST Tag Recombinant Protein
628.00 € - 7890.00 €
Specifications
Accession Number | NP_005618, S589D |
---|---|
Gene Alias | SGK (SGK1) |
KinaseFamily | SGK Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15418573
|
Invitrogen™
PR7358A |
100μg |
1826.00 €
100µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11503557
|
Invitrogen™
PV3818 |
10 μg |
628.00 €
10µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10392363
|
Invitrogen™
PV4056 |
1mg |
7890.00 €
1mg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SGK (SGK1) may be a transcriptional target for p38/MAPK in hepatoma cells.
Specifications
NP_005618, S589D | |
SGK Kinase Family | |
Recombinant | |
Baculovirus |
SGK (SGK1) | |
Human | |
GST Tag |