Learn More
SGE™ Terry Tool Tubing Cutter Replacement Part
Product Code. 10507162
Change view
Click to view available options
Quantity:
3/Pk.
Unit Size:
Pack of 3
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10507162 3/Pk. Pack of 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10507162 Supplier SGE™ Supplier No. 082781
Only null left
Add to basket
Add to basket
Replacement cutting wheel for use with Terry-Tool Tubing Center.
  • Replacement wheel
  • For cutting 0.0625 in. and 0.125 in. stainless steel tubing
Specifications
Description Replacement cutting wheel
For Use With (Application) Terry-Tool Tubing Center
Quantity 3/Pk.