Learn More
SGE™ Replacement Needles for Removable Needle Syringes
Click to view available options
Needle Gauge:
23 G
25 G
Diameter (English) Outer Needle:
0.019 in.
0.024 in.
Unit Size:
Pack of 2
Pack of 5
Description
- For 10μL syringes
- 50mm (2 in.) exposed length when fitted to syringe
Specifications
Specifications
| Needle Gauge | 25 G |
| Diameter (English) Outer Needle | 0.019 in. |
| Diameter (Metric) Outer | 0.5 mm |
| Diameter (Metric) Outer Needle | 0.5 mm |
| For Use With (Application) | Gas chromatography |
| Length (English) | 2 in. |
| Length (English) Needle | 1.9 in. |
| Length (Metric) | 50 mm |
| Product Type | Replacement Needle |
| Quantity | 2/Pk. |
| Show More |