Learn More
Trajan™ PEEKsil™ Tubing
Mechanically robust polymer-sheathed fused silica tubing. Trajan Scientific and Medical™ PEEKsil™ Tubing can be used for sealing with conventional metal or polymer ferrule systems.
65.50 € - 117.00 €
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Overall Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Diameter (Metric) Outer | Overall Length (Metric) | Price | Quantity & Availability | |||
15238753
|
Trajan™
0624251 |
0.05 mm | 1.5 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15298773
|
Trajan™
0624350 |
0.175 mm | 1.5 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15248753
|
Trajan™
0624252 |
0.05 mm | 1.5 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15208783
|
Trajan™
0624351 |
0.175 mm | 1.5 mm | 50 mm |
96.80 €
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15218783
|
Trajan™
0624352 |
0.175 mm | 1.5 mm | 100 mm |
104.00 €
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15228783
|
Trajan™
0624353 |
0.175 mm | 1.5 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15228753
|
Trajan™
0624250 |
0.05 mm | 1.6 mm | 500 mm |
65.50 €
Pack of 2 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
15258753
|
Trajan™
0624253 |
0.05 mm | 1.5 mm | 200 mm |
117.00 €
Pack of 5 |
Estimated Shipment: 03-06-2024
Log in to see stock. |
|||
Description
- Robust - PEEK polymer exterior and fused silica inner tubing
- Smooth wall surface giving excellent flow characteristics
- Compatible with most organic solvents
- Inertness - extremely low absorption characteristics
- Precise ID
Specifications
Tubing |