Learn More
Trajan™ SilTite™ GC Capillary Column Connectors: μ Unions
Product Code. 15278933
Click to view available options
Connections:
0.1-0.25 mm and 0.1-0.25 mm
0.1-0.25 mm and 0.32 mm
0.1-0.25 mm and 0.53 mm
0.32 mm and 0.32 mm
0.32 mm and 0.53 mm
0.53 mm and 0.53 mm
Unit Size:
Each
Product Code. 15278933
Brand: Trajan™ 073565
Only null left
Add to basket
Add to basket
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
  • Low thermal mass
  • Zero Dead Volume
  • Inert
  • No-need to retighten after installation
  • Connect Pre-column or GC x GC
Specifications
Type Connector
Connections 0.53 mm and 0.53 mm
Includes 2 unions, 5 double taper ferrules and tools
For Use With (Equipment) GC Capillary Column