Learn More
SGE™ BP20 (Wax) GC Capillary Columns
Combines innovative phase technology and unique fused silica production capability. SGE™ BP20 (Wax) GC Capillary Columns provides end-to-end separation solutions for all applications.
Brand:  SGE™ 054440
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15288673
  • 835.00 € / Each
Estimated Shipment: 20-06-2024
to see stock.
Add to basket
Description
Description
Features
  • Industry standard Wax column
  • Bonded and cross-linked
  • Can be solvent rinsed
Applications
  • Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications
Specifications
0.53 mm
Hydrogen Bonding or Dipole interactions
280°C
20°C to 260/280°C
30 m
Fused Silica
G14, G15, G16, G20, G39
SGE™ BP20 (Wax)
FAMEs Analysis, Free Acids, Fatty Acid Methyl Esters, Aromatics Analysis, Solvents and Essential Oils
0.5 μm
Polar