Learn More
SGE™ ConnecTite™ Liners for Thermo Scientific™ GC
Designed for use with Thermo Scientific TRACE™ and FOCUS™ GCs
Brand:  SGE 092334
 View more versions of this product
Product Code. 15239023
  • 376.00 € / Pack of 5
Estimated Shipment: 26-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Range covers all applications and all popular instrument makes/ models
  • Range covers vaporizing and cool on-column injection techniques
  • Inlet liners ensure complete transfer of sample from syringe to the column without loss
  • 8mm OD, 5mm ID; 105mm length
Specifications
Specifications
5/Pk.
Connectite Standard Liner
0.8 cm
10.5 cm
0.5 cm
Thermo Scientific GC Trace and Focus