Learn More
Invitrogen™ SF3A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530267
Description
Recommended positive controls: HeLa. Predicted reactivity: Mouse (99%), Rat (98%), Zebrafish (95%), Xenopus laevis (98%), Chicken (98%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes subunit 2 of the splicing factor 3a protein complex. The splicing factor 3a heterotrimer includes subunits 1, 2 and 3 and is necessary for the in vitro conversion of 15S U2 snRNP into an active 17S particle that performs pre-mRNA splicing. Subunit 2 interacts with subunit 1 through its amino-terminus while the single zinc finger domain of subunit 2 plays a role in its binding to the 15S U2 snRNP. Subunit 2 may also function independently of its RNA splicing function as a microtubule-binding protein.
Specifications
| SF3A2 | |
| Polyclonal | |
| Unconjugated | |
| SF3A2 | |
| 66kDa; AW047242; pre-mRNA splicing factor SF3A, subunit 2; PRP11; PRPF11; SAP 62; Sap62; Sf3a2; SF3a66; SFA66; spliceosomal protein gene 62; spliceosome associated protein 62; spliceosome-associated protein 62; Splicing factor 3A subunit 2; splicing factor 3a, subunit 2; splicing factor 3a, subunit 2, 66kD; splicing factor 3a, subunit 2, 66kDa | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8175 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.33 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q15428 | |
| SF3A2 | |
| Recombinant protein encompassing a sequence within the center region of human SAP62. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.