missing translation for 'onlineSavingsMsg'
Learn More

SF2523, MedChemExpress

Product Code. 16452706
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16452706

Brand: Medchem Express HY101146

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress SF2523 is a highly selective and potent inhibitor of PI3K with IC50s of 34 nM, 158 nM, 9 nM, 241 nM and 280 nM for PI3Kα, PI3Kγ, DNA-PK, BRD4 and mTOR, respectively.

SF2523 is a highly selective and potent inhibitor of PI3K with IC50s of 34 nM, 158 nM, 9 nM, 241 nM and 280 nM for PI3Kα, PI3Kγ, DNA-PK, BRD4 and mTOR, respectively.

Specifications

Chemical Name or Material SF2523
CAS 1174428-47-7
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C19H17NO5S
Quantity 25 mg
Solubility Information DMSO : ≥ 30 mg/mL (80.77 mM)
SMILES O=C1C=C(N2CCOCC2)OC3=C1SC=C3C4=CC=C(OCCO5)C5=C4
Molecular Weight (g/mol) 371.41
Formula Weight 371.41
Percent Purity 97.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.