Learn More
Seryl-tRNA synthetase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529761
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: 293T, HeLa, HepG2. Predicted reactivity: Mouse (96%), Rat (94%), Zebrafish (81%), Xenopus laevis (84%), Chicken (87%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Seryl-tRNA synthetase (SerRS) catalyzes the attachment of serine to tRNA(Ser) in a two-step reaction: serine is first activated by ATP to form Ser-AMP and then transferred to the acceptor end of tRNA(Ser). Is also probably able to aminoacylate tRNA(Sec) with serine, to form the misacylated tRNA L-seryl-tRNA(Sec), which will be further converted into selenocysteinyl-tRNA(Sec).Specifications
Seryl-tRNA synthetase | |
Polyclonal | |
Unconjugated | |
SARS1 | |
EC 6.1.1.11; SARS; Sars1; serine tRNA ligase 1, cytoplasmic; serine--tRNA ligase, cytoplasmic; SerRS; SERS; seryl-aminoacyl-tRNA synthetase; seryl-aminoacyl-tRNA synthetase 1; seryl-tRNA synthetase; seryl-tRNA synthetase, cytoplasmic; seryl-tRNA(Ser/Sec) synthetase; Strs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20226, 266975, 6301 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.51 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
P26638, P49591, Q6P799 | |
SARS1 | |
Recombinant fragment corresponding to a region within amino acids 28 and 372 of Human Seryl-tRNA synthetase | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |