missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Seryl tRNA synthetase Polyclonal antibody specifically detects Seryl tRNA synthetase in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | Seryl tRNA synthetase |
| Applications | ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | HRP |
| Formulation | PBS |
| Gene Alias | EC 6.1.1.11, FLJ36399, serine-tRNA ligase, Serine--tRNA ligase, SERRS, SERSserine tRNA ligase 1, cytoplasmic, Seryl-tRNA Ser/Sec synthetase, seryl-tRNA synthetase, seryl-tRNA synthetase, cytoplasmic, Seryl-tRNA(Ser/Sec) synthetase |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human Seryl tRNA synthetase (NP_006504.2).,, Sequence:, MVLDLDLFRVDKGGDPALIRETQEKRFKDPGLVDQLVKADSEWRRCRFRADNLNKLKNLCSKTIGEKMKKKEPVGDDESVPENVLSFDDLTADALANLKVSQIKKVRLLIDEAILKCDAERIKLEAERFENLREIGNLLHPSVPISNDEDVDNKVERIWGDCTVRKKYSHVDLVVMVDGFEGEKGAVVAGSRGYFLKGVLVFLEQALIQYALRTLGSRGYIPIYTPFFMRKEVMQEVAQLSQFDEELYKVIGKGSEKSDDNSYDEKYLIATSEQPIAALHRDEWLRPEDLPIKYAGLSTC |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?