Learn More
BD Vacutainer Serum Tubes
Product Code. 12841863
Change view
Click to view available options
Capacity (Metric):
4 mL
Closure Color:
Red
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color unitSize
12841863 4 mL Red Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12841863 Supplier BD Vacutainer Supplier No. 369032
Please to purchase this item. Need a web account? Register with us today!
Plastic (PET) tubes for serum collection.
  • Silica particles added as the clot activator
  • Labelled with CAT (clot activator tube)
  • Should be gently inverted 180° and back 5-6 times
TRUSTED_SUSTAINABILITY
Specifications
Additive Silica (Clot Activator)
Material PET
Closure Type Hemogard
Capacity (Metric) 4 mL
Closure Color Red
Tube Size 13 x 75 mL
Label Type Paper
compliance-icons
Product Identifier
  • TUBE PLN PLH 13X75 4.0 PLBLCE RD SIL
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Carcinogenicity Category 1A
Hazard Statement
  • H350-May cause cancer.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: quartz