Learn More
Sertindole, MedChemExpress
Product Code. 16466088
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Product Code. 16466088
Brand: Medchem Express HY14543
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Sertindole, a neuroleptic, is one of the newer antipsychotic medications available.
Sertindole, a neuroleptic, is one of the newer antipsychotic medications available.
Specifications
Chemical Name or Material Sertindole
CAS 106516-24-9
Color White
Purity Grade Notes Research
Molecular Formula C24H26ClFN4O
Quantity 10 mg
Synonym Lu 23-174
Solubility Information DMSO : 25 mg/mL (56.70 mM; Need ultrasonic)
SMILES FC1=CC=C(N2C3=CC=C(Cl)C=C3C(C4CCN(CCN5CCNC5=O)CC4)=C2)C=C1
Molecular Weight (g/mol) 440.94
Formula Weight 440.94
Percent Purity 95.04%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation