missing translation for 'onlineSavingsMsg'
Learn More

Sertaconazole nitrate, MedChemExpress

Product Code. 16434467
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16434467 50 mg 50mg
16458436 100 mg 100mg
16428598 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16434467 Supplier Medchem Express Supplier No. HYB0736A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent, exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway. Sertaconazole nitrate is also a microtubule inhibitor, shows antiproliferative effect, induces apoptosis and autophagy, and can also inhibit the migration of cells.

Sertaconazole nitrate (FI7056) is a broad-spectrum topical antifungal agent, exhibits anti-inflammatory activity via activation of a p38-COX-2-PGE2 pathway. Sertaconazole nitrate is also a microtubule inhibitor, shows antiproliferative effect, induces apoptosis and autophagy, and can also inhibit the migration of cells.

Specifications

Chemical Name or Material Sertaconazole nitrate
Physical Form Solid
CAS 99592-39-9
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C20H16Cl3N3O4S
For Use With (Application) COVID-19-immunoregulation
Synonym FI7056
Solubility Information DMSO : ≥ 100 mg/mL (199.69 mM)
SMILES ClC1=CC=C(C(OCC2=CSC3=C(Cl)C=CC=C32)CN4C=CN=C4)C(Cl)=C1.O[N+]([O-])=O
Molecular Weight (g/mol) 500.78
Formula Weight 500.78
Percent Purity 99.58%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.