missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ SERPINI2 (Human) Recombinant Protein
Human SERPINI2 full-length ORF ( NP_006208, 1 a.a. - 405 a.a.) recombinant protein with GST-tag at N-terminal.
335.00 € - 508.00 €
Specifications
Accession Number | NP_006208 |
---|---|
Gene ID (Entrez) | 5276 |
Name | serpin peptidase inhibitor, clade I (pancpin), member 2 |
Preparation Method | Wheat germ expression system |
Quality Control Testing | 125% SDS-PAGE Stained with Coomassie Blue |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16172775
|
Abnova™
H00005276-P01.10UG |
10 μg |
335.00 €
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16182775
|
Abnova™
H00005276-P01.25UG |
25 μg |
508.00 €
25µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Sequence: MDTIFLWSLLLLFFGSQASRCSAQKNTEFAVDLYQEVSLSHKDNIIFSPLGITLVLEMVQLGAKGKAQQQIRQTLKQQETSAGEEFFVLKSFFSAISEKKQEFTFNLANALYLQEGFTVKEQYLHGNKEFFQSAIKLVDFQDAKACAEMISTWVERKTDGKIKDMFSGEEFGPLTRLVLVNAIYFKGDWKQKFRKEDTQLINFTKKNGSTVKIPMMKALLRTKYGYFSESSLNYQVLELSYKGDEFSLIIILPAEGMDIEEVEKLITAQQILKWLSEMQEEEVEISLPRFKVEQKVDFKDVLYSLNITEIFSGGCDLSGITDSSEVYVSQVTQKVFFEINEDGSEAATSTGIHIPVIMSLAQSQFIANHPFLFIMKHNPTESILFMGRVTNPDTQEIKGRDLDSL
Specifications
NP_006208 | |
serpin peptidase inhibitor, clade I (pancpin), member 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
MEPI, PANCPIN, PI14, TSA2004 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |
5276 | |
Wheat germ expression system | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SERPINI2 | |
GST |