missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SENP1 Polyclonal Antibody

Product Code. 10631234
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10631234 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Product Code. 10631234 Supplier Invitrogen™ Supplier No. 383350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Research Use Only. All usage must comply with product instructions.

Rabbit Polyclonal Antibody

SENP1 Polyclonal Antibody for Western Blot, ICC/IF, IP

SENP1 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SENP deconjugates SUMO1 from HIPK2 and from HDAC1, which decreases the transcriptional repression activity of the latter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SENP1
Applications Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene SENP1
Gene Accession No. Q9P0U3
Gene Alias 2310046A20Rik; D15Ertd528e; E330036L07Rik; SEN1; Senp1; sentrin/SUMO-specific protease SENP1; Sentrin-specific protease 1; SUMO-1 protease 2; SUMO1/sentrin specific peptidase 1; SUMO1/sentrin specific protease 1; Sumo1/sentrin/SMT3 specific peptidase 1; Supr2; suPr-2
Gene Symbols SENP1
Host Species Rabbit
Immunogen Synthetic peptide derived from the mid region of the human SENP1 protein.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29843
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.