missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Advantage-Lab™ Semi-Automatic Vertical Autoclave
Click to view available options
Capacity (Metric):
23 L
62 L
85 L
Unit Size:
Each
Description
- Suitable for the sterilization of the following:
- Liquids such as nutrient media and buffer solutions.
- Solid items such as pipettes, tubes and filters.
- Glassware and plastic articles.
- Waste sterilization.
- Temperature and pressure are controlled through sensors.
- Temperature sensor on a flexible cable that can be used in chamber as well as inside liquid or media reference bottles.
- Pressure dependent door lock that prevents door opening when under pressure.
- Six individual programs available.
- Sterilization Parameters:
- Temperature and time are freely selectable.
- Temperature can be set between 60 and 137°C.
- Sterilization time can be set from 1-99 minutes.
- Pre-selected parameters remain programmed until altered.
- Temperature dependent door lock works in conjunction with temperature control in a reference bottle so that the system prevents the door from being opened until the temperature is below a specified level, preventing delayed boiling.
- Water level safety device that prevents the autoclave from being heated if insufficient water is available for steam generation.
- Display of temperature, pressure and time with a visible and acoustic alarm.
Specifications
Specifications
| Controller Type | Semi-automatic electro-mechanical control |
| Color | Cream White, Stainless Steel |
| Capacity (Metric) | 85 L |
| Model | Floor-standing |
| Product Type | Autoclave, Vertical |
| Temperature (Metric) | 100°C to 134°C |
| Weight (Metric) | 125 kg |
| Material | Stainless Steel |
| Operating Pressure | 0 to 230 kPa |
| Voltage | 380/400 V |
| Show More |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction
