missing translation for 'onlineSavingsMsg'
Learn More

Advantage-Lab™ Semi-Automatic Vertical Autoclave

Product Code. 10695033
Encompass
Click to view available options
Capacity (Metric):
23 L
62 L
85 L
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10695033

Brand: Advantage-Lab™ AL0210100

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Sterilize loads with low sensitivity. Advantage-Lab™ Semi-Automatic Vertical Autoclave is designed to be placed on the floor.

  • Suitable for the sterilization of the following:
    • Liquids such as nutrient media and buffer solutions.
    • Solid items such as pipettes, tubes and filters.
    • Glassware and plastic articles.
    • Waste sterilization.
  • Temperature and pressure are controlled through sensors.
  • Temperature sensor on a flexible cable that can be used in chamber as well as inside liquid or media reference bottles.
  • Pressure dependent door lock that prevents door opening when under pressure.
  • Six individual programs available.
  • Sterilization Parameters:
    • Temperature and time are freely selectable.
    • Temperature can be set between 60 and 137°C.
    • Sterilization time can be set from 1-99 minutes.
    • Pre-selected parameters remain programmed until altered.
  • Temperature dependent door lock works in conjunction with temperature control in a reference bottle so that the system prevents the door from being opened until the temperature is below a specified level, preventing delayed boiling.
  • Water level safety device that prevents the autoclave from being heated if insufficient water is available for steam generation.
  • Display of temperature, pressure and time with a visible and acoustic alarm.

Specifications

Controller Type Semi-automatic electro-mechanical control
Color Cream White, Stainless Steel
Capacity (Metric) 85 L
Model Floor-standing
Product Type Autoclave, Vertical
Temperature (Metric) 100°C to 134°C
Weight (Metric) 125 kg
Material Stainless Steel
Operating Pressure 0 to 230 kPa
Voltage 380/400 V
Dimensions (L x W x H) 500 x 650 x 950 mm
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.