missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Semaglutide TFA, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
Description
Semaglutide TFA, a long-acting GLP-1 analogue, is a glucagon-like peptide-1 (GLP-1) receptor agonist. Semaglutide TFA has the potential for type 2 diabetes treatment.
Specifications
Specifications
| Chemical Name or Material | Semaglutide TFA |
| Physical Form | Solid |
| Color | White |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C189H292F3N45O61 |
| For Use With (Application) | Metabolism-sugar/lipid metabolism |
| Solubility Information | H2O : 100 mg/mL (23.65 mM; Need ultrasonic) |
| SMILES | [Semaglutide (TFA salt)] |
| Molecular Weight (g/mol) | 4227.66 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction