Learn More
Brady™ Self-Laminating Rotating Vinyl Wrap Around Wire and Cable Labels for M6/M7 Printers
Product Code. 17838164
Change view
Click to view available options
Dimensions (L x W):
1.2 x 1 in.
1.75 x 0.5 in.
1.75 x 1 in.
2.75 x 0.5 in.
2.75 x 1 in.
3.125 x 1 in.
3.125 x 1.5 in.
Height (Metric):
30.48 mm
44.45 mm
69.85 mm
79.38 mm
Unit Size:
Pack of 100
Pack of 50
Pack of 75
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Height (Metric) unitSize
17838164 1.75 x 0.5 in. 44.45 mm Pack of 75
17868474 1.2 x 1 in. 30.48 mm Pack of 100
17828194 1.75 x 1 in. 44.45 mm Pack of 75
17818314 2.75 x 0.5 in. 69.85 mm Pack of 50
17828164 2.75 x 1 in. 69.85 mm Pack of 50
17878244 3.125 x 1 in. 79.38 mm Pack of 50
17848384 3.125 x 1.5 in. 79.38 mm Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 17838164 Supplier Brady™ Supplier No. 174167
  • ONLINE EXCLUSIVE PRICE
    52.15 €
    47.43 € / Pack of 75
    Save 4.72 €
    9% Off
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
Identify your wires from any angle with Rotating Vinyl Wire Labels. Simply print your labels and then peel, fold and wrap – they’ll rotate freely for easy ID on even closed network connections.
  • Rotates so you can identify your labels from any angle
  • Simply wrap and apply--there's no adhesive residue or label parts to cling to your wires
  • Spins freely without anchors so these labels won't damage your Cat 5 / 6 and fiber optic cables
  • Made of B-427 vinyl
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance ASTM D 1000, ASTM D 2979, ASTM G 155, UL Recognized
Adhesive Type Permanent Acrylic
Coating None
Color Clear, White
Finish Matte
Format Die-cut
Material Vinyl
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Label
Sterility Non-sterile
For Use With (Application) Data and Telecommunications Labeling, Electrical Labeling, Laboratory Labeling, Wire and Cable Labeling
For Use With (Equipment) M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Latex Free No
Quantity 75 Labels
Shape Rectangle
Temperature Range (English) -94°F to +158°F
Temperature Range (Metric) -70°C to +70°C
Thickness (English) 0.004 in.
Thickness (Metric) 0.102 mm
Dimensions (L x W) 1.75 x 0.5 in.
Length (English) 1.75 in.
Width (English) 0.5 in.
Width (Metric) 12.7 mm
Width (English) Printable Area 0.5 in.
Width (Metric) Printable Area 12.7 mm
Height (English) Printable Area 0.7 in.
Height (Metric) 44.45 mm
Height (Metric) Printable Area 17.78 mm
Alternate Part Number M6-206-427-RO