Learn More
Thermo Scientific™ Selenite Cystine Broth
Product Code. 13285799
Change view
Click to view available options
Description:
Selenite Cystine Broth
Quantity:
50/Pk.
Fill Volume:
10 mL
Fill Weight:
10 ± 0.5mL
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity Fill Volume Fill Weight Unit Size
13285799 Selenite Cystine Broth 50/Pk. 10 mL 10 ± 0.5mL Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13285799 Supplier Thermo Scientific™ Supplier No. TV5018E
  • ONLINE EXCLUSIVE PRICE
    82.00 €
    53.98 € / Pack of 50
    Save 28.02 €
    34% Off
 Restricted Item
Estimated Shipment: 06-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Enrich for Salmonella species from feces or food samples with Thermo Scientitfic™ Selenite Cystine Broth prepared media tubes.
Enrich for Salmonella species from feces or food samples with Thermo Scientitfic™ Selenite Cystine Broth. The formulation corresponds to that recommended by the AOAC1 for detection of Salmonella in foodstuffs, in particular, egg products. It is included among the standard methods media of the American Public Health Association2, 3. It also complies with the requirements of the United States Pharmacopoeia4.
Selenite Cystine Broth is modified from the formula of Leifson5 with added cystine6. This addition has given favorable results in many studies7.
The effect of the cystine may be due to its reducing abilities, which will lower the toxicity of selenite to microorganisms, and/or the extra organic sulfur provided may have a sparing effect on the critical sulfur components of the bacteria, again reducing the selective effect of the selenite.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Color Light ivory, transparent
Content And Storage 2°C to 12°C
Description Selenite Cystine Broth
Format Tube
Manufacturing Origin Germany
Product Type Microbiology Media
Fill Volume 10 mL
Fill Weight 10 ± 0.5mL
Packaging Type 50 tubes in a carton box
Quantity 50/Pk.
For In Vitro Diagnostic Use.