missing translation for 'onlineSavingsMsg'
Learn More

Carl Roth™ Nappa Leather Safety Gloves

Product Code. 10496393
Encompass
Click to view available options
Size:
10
This item is not returnable. View return policy

Product Code. 10496393

Brand: Carl Roth™ 1748.1

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Soft, specially selected Nappa leather (goat or sheep) gloves with leather finger walls.

  • Gloves offer moderate protection against mechanical hazards according to EN 420, EN 388 (2111)

Specifications

Certifications/Compliance EN 420
Certifications/Compliance (Europe) EN 420, EN 388 (2111)
Color White
Hand Specific
Material Nappa Leather
Product Type Safety Gloves
Size 10
For Use With (Application) For Hot and Cold, Comfortable to Wear, Due to Tear and Water Resistant
Quantity 1 Pack
Resistant To Tear and Water Resistant
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.