Learn More
3M™ Secure Click™ Filter Retainer D701
3M™ Secure Click™ Filter Retainer D701 is designed for use with 3M™ Secure Click™ Respirators 800 Series.
Brand:  3M™ 7100184498
220.00 EUR valid until 2024-12-31
Use promo code "22912" to get your promotional price.
Additional Details : Weight : 0.21000kg
Product Code. 16496370
  • 341.00 € / 50 pairs
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
Description
Description
  • Used to hold 3M™ Secure Click™ Particulate Filters D7N11 or D7P71 in place on 3M™ Secure Click™ Cartridges D8000 Series
Specifications
Specifications
3M™ Secure Click™ HF-800 Series Reusable Respirators
100/Cs.