missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seco Rapamycin sodium salt, MedChemExpress
Description
Seco Rapamycin sodium salt is the ring-opened product of Rapamycin. Seco-rapamycin is reported not to affect the mTOR function.
Specifications
Specifications
| Chemical Name or Material | Seco Rapamycin sodium salt |
| CAS | 148554-65-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C51H78NNaO13 |
| Quantity | 5 mg |
| Synonym | Secorapamycin A monosodium |
| Solubility Information | DMSO : ≥ 46 mg/mL (49.14 mM) |
| SMILES | O=C([C@H]1N(C(C([C@@]2(O)[C@H](C)CC[C@@H](C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C([C@H](OC)[C@H](O)/C(C)=C/[C@@H](C)C(/C=C/[C@H](C)C[C@H]3C[C@@H](OC)[C@H](O)CC3)=O)=O)O2)=O)=O)CCCC1)O[Na] |
| Molecular Weight (g/mol) | 936.15 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?