Learn More
Seco Rapamycin sodium salt, MedChemExpress
Product Code. 16480268
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16480268 5 mg 5mg
16447808 10 mg 10mg
16443656 25 mg 25mg
16403767 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16480268 Supplier Medchem Express Supplier No. HY19555A
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Seco Rapamycin sodium salt is the ring-opened product of Rapamycin. Seco-rapamycin is reported not to affect the mTOR function.
Seco Rapamycin sodium salt is the ring-opened product of Rapamycin. Seco-rapamycin is reported not to affect the mTOR function.
Specifications
Chemical Name or Material Seco Rapamycin sodium salt
CAS 148554-65-8
Color White
Purity Grade Notes Research
Molecular Formula C51H78NNaO13
Quantity 5 mg
Synonym Secorapamycin A monosodium
Solubility Information DMSO : ≥ 46 mg/mL (49.14 mM)
SMILES O=C([C@H]1N(C(C([C@@]2(O)[C@H](C)CC[C@@H](C[C@H](OC)/C(C)=C/C=C/C=C/[C@@H](C)C[C@@H](C)C([C@H](OC)[C@H](O)/C(C)=C/[C@@H](C)C(/C=C/[C@H](C)C[C@H]3C[C@@H](OC)[C@H](O)CC3)=O)=O)O2)=O)=O)CCCC1)O[Na]
Molecular Weight (g/mol) 936.15
Formula Weight 936.15
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation