Learn More
Secnidazole, MedChemExpress
Product Code. 16468668
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
500 mg
Unit Size:
100mg
10mg
500mg
50mg
Product Code. 16468668
Brand: Medchem Express HYB1118
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Secnidazole (RP-14539) is an orally active azole antibiotic and a imidazole mitigator of Serratia marcescens virulence.
Secnidazole, as an analog of acylhomoserine lactones, effectively inhibits QS resulting in the attenuation of Pseudomonas aeruginosa pathogenesis. Secnidazole has antimicrobial activity against many anaerobic Gram-negative and Gram-positive bacterial species in vitro. Secnidazole can be used for the research of various diseases, such as amoebiasis and giardiasis, and bacterial vaginitis.
Specifications
Chemical Name or Material Secnidazole
Physical Form Solid
CAS 3366-95-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C7H11N3O3
For Use With (Application) COVID-19-immunoregulation
Synonym RP-14539 PM-185184
Solubility Information DMSO : ≥ 30 mg/mL (162.00 mM) ∣H2O : 25 mg/mL (135.00 mM; Need ultrasonic)
SMILES OC(C)CN1C([N+]([O-])=O)=CN=C1C
Molecular Weight (g/mol) 185.18
Formula Weight 185.18
Percent Purity 99.88%
Grade Research