Learn More
SDHB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10620-1-AP-20UL
 View more versions of this product
Product Code. 16837113
  • 145.00 € / 20µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Complex II of the respiratory chain, which is specifically involved in the oxidation of succinate, carries electrons from FADH to CoQ. The complex is composed of four nuclear-encoded subunits and is localized in the mitochondrial inner membrane. The iron-sulfur subunit is highly conserved and contains three cysteine-rich clusters which may comprise the iron-sulfur centers of the enzyme. Sporadic and familial mutations in this gene result in paragangliomas and pheochromocytoma, and support a link between mitochondrial dysfunction and tumorigenesis.
Specifications
Specifications
SDHB
Polyclonal
Unconjugated
SDHB
IP, PGL4, SDH, SDH1, SDHB, SDHIP
Rabbit
Antigen Affinity Chromatography
RUO
298596, 6390, 67680
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P21912, P21913, Q9CQA3
SDHB
SDHB Fusion Protein Ag0968
20 μL
Primary
Human, Rat, Mouse