Learn More
Scutellarin, MedChemExpress
Product Code. 16453896
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16453896 25 mg 25mg
16419088 10 mg 10mg
16404887 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16453896 Supplier Medchem Express Supplier No. HYN0751
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Scutellarin, an active flavone isolated from Scutellaria baicalensis, can down-regulates the STAT3/Girdin/Akt signaling in HCC cells, and inhibits RANKL-mediated MAPK and NF-κB signaling pathway in osteoclasts. Scutellarin is active against HIV-1IIIB, HIV-1(74V) and HIV-1KM018 with EC50s of 26 μM, 253 μM and 136 μM, respectively.
Scutellarin, an active flavone isolated from Scutellaria baicalensis, can down-regulates the STAT3/Girdin/Akt signaling in HCC cells, and inhibits RANKL-mediated MAPK and NF-κB signaling pathway in osteoclasts. Scutellarin is active against HIV-1IIIB, HIV-1(74V) and HIV-1KM018 with EC50s of 26 μM, 253 μM and 136 μM, respectively.
Specifications
Chemical Name or Material Scutellarin
CAS 27740-01-8
Color Light Green
Purity Grade Notes Research
Molecular Formula C21H18O12
Quantity 25 mg
Solubility Information DMSO : 100 mg/mL (216.28 mM; Need ultrasonic)
SMILES O=C(C=C(C1=CC=C(O)C=C1)OC2=CC(O[C@@H]([C@@H]([C@@H](O)[C@@H]3O)O)O[C@@H]3C(O)=O)=C4O)C2=C4O
Molecular Weight (g/mol) 462.36
Formula Weight 462.36
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease