Learn More
Thermo Scientific Chemicals Scutellarin
Product Code. 16524474 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16524474 250 mg 250mg
16514474 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16524474 Supplier Thermo Scientific Chemicals Supplier No. 465132500
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 27740-01-8 | C21H18O12 | 462.36 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 27740-01-8
Molecular Weight (g/mol) 462.36
InChI Key DJSISFGPUUYILV-ZFORQUDYSA-N
IUPAC Name (2S,3S,4S,5R,6S)-6-{[5,6-dihydroxy-2-(4-hydroxyphenyl)-4-oxo-4H-chromen-7-yl]oxy}-3,4,5-trihydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@H](OC2=C(O)C(O)=C3C(=O)C=C(OC3=C2)C2=CC=C(O)C=C2)O[C@@H]([C@H]1O)C(O)=O
Specifications
Molecular Formula C21H18O12
Quantity 250 mg
Formula Weight 462.36
Chemical Name or Material Scutellarin